Learn More
PTS Diagnostics PTS PANELS™ HDL Cholesterol Test Strips
Quick and easy measurement of HDL Cholesterol from whole blood using CardioChek™ PA Analyzer for professional and home use
Supplier: PTS Diagnostics 1714
Includes: PTS PANELS HDL Cholesterol Test Strips, MEMo Chip (contains lot-specific test strip information), Instructions
Requires : (Not provided)
CardioChek brand analyzer, Quality Control Materials, Lancets for fingerstick (or venous blood collection supplies), Alcohol wipes and/or gauze, Capillary Blood Collector or other precision pipet for blood collection and application
Description
PTS PANELS HDL Cholesterol Test strips provide a quantitative measurement of High Density Lipoprotein (HDL) Cholesterol. This system is intended to assist in screening for decreased HDL cholesterol, a risk factor in coronary heart disease. HDL Cholesterol, often referred to as “good cholesterol”, is an important component of a lipid profile. A low HDL indicates increased risk of coronary heart disease, while a high HDL indicates decreased risk.
- A MEMo Chip™ is provided with each package of test strips and must be properly inserted into the analyzer before any test can be run
- The MEMo Chip contains test name,calibration curve, lot number and test strip expiration date
- After the test strip is inserted into the analyzer and blood applied to the strip, test results are displayed in about a minute
Individuals should consult their healthcare professional with any questions about HDL levels and when to use this test.
Specifications
Waived | |
Total: HDL Cholesterol 25 to 85mg/dL (0.65 to 2.20mM/L) | |
2 min. |
25 Pack | |
15 μL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.