Learn More
Aviva Systems Biology Corp PTTG2 Antibody - middle region (ARP50542_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50542_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTTG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PTTG2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-60374