Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTTG2 Mouse anti-Human, Purified MaxPab™ Polyclonal , Abnova™
Mouse polyclonal antibody raised against a full-length human PTTG2 protein.
Supplier: Abnova Corporation H00010744B01P
Description
Sequence: MATLIYVDKEIGEPGTRVAAKDVLKLESRPSIKALDGISQVLTPRFGKTYDAPSALPKATRKALGTVNRATEKSVKTNGPRKQKQPSFSAKKMTEKTVKTKSSVPASDDAYPEIEKFFPFNLLDFESFDLPEERQIAHLPLSGVPLMILDEEGELEKLFQLGPPSPVKMPSPPWECNLFAVSFKHSVDPGCSpecifications
PTTG2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human PTTG2 protein. | |
PTTG2 | |
PTTG2 | |
PTTG2 (NP_006598.1,1 a.a. ∽ 191 a.a) full-length human protein. | |
50 μg | |
Primary | |
Antibody reactive against mammalian transfected lysate. | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_006607 | |
Mouse | |
Protein A/G purification | |
RUO | |
10744 | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction