Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTX3 Mouse anti-Mouse, Clone: 6B11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA533377

Catalog No. PIMA533377


Add to cart

Description

Description

Product is supplied at 1 mL (100 μL/mL). Application Notes: Western blot - A reduced sample treatment and SDS-Page was used. Positive control: Section of inflamed tissues; Negative control: Section of normal tissues.

Pentraxins are a protein superfamily that is characterized by a cyclic multimeric structure. Ptx3, also known as tumor necrosis factor-stimulated gene sequence-14 (TSG14), is a secreted pattern-recognition receptor that has a non-redundant role in resistance to selected microbial agents. Ptx3 belongs to the family of "long pentraxins", which have C-terminal pentraxin domains and novel amino-terminal domains. Ptx3 binds selected pathogens, including Aspergillus fumigatus, Pseudomonas aeruginosa and Salmonella typhimurium. It is synthesized in IgA glomerulonephritis and activates mesangial cells. Secretion of Ptx3 in adipose cells can be induced by TNF . Ptx3 is also involved in amplification of inflammatory reactions and regulation of innate immunity. The human PTX3 gene maps to chromosome 3q25.
Specifications

Specifications

PTX3
Monoclonal
100 μg/mL
PBS with 0.1% BSA and no preservative
P48759
Pentraxin-related protein PTX3, Pentaxin-related protein PTX3, Tumor necrosis factor alpha-induced protein 5, TNF alpha-induced protein 5, Tumor necrosis factor-inducible gene 14 protein, TSG-14, TNFAIP5, TSG14, PTX3
Mouse
IgG1
100 μg
4° C
Primary
19288
ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Western Blot
6B11
Unconjugated
Ptx3
liquid
Ptx3
Recombit mouse PTX3
Protein G
RUO
Antibody
Monoclonal
Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit