Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PU 02, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
313984-77-9
C16H12N4S
292.36
BGMSTNYJYPSLHN-UHFFFAOYSA-N
6-1-naphthalenylmethyl thio-9h-purine,6-naphthalen-1-ylmethylsulfanyl-7h-purine,6-naphthalen-1-ylmethyl sulfanyl-9h-purine,6-naphthylmethylthio purine,6-1-naphthylmethyl sulfanyl-9h-purine,6-naphthalen-1-ylmethyl thio-7h-purine,6-naphthalen-1-ylmethylsulfanyl-9h-purine
720937
6-(naphthalen-1-ylmethylsulfanyl)-7H-purine
C1=CC=C2C(=C1)C=CC=C2CSC3=NC=NC4=C3NC=N4
This item is not returnable. View return policy
313984-77-9
C16H12N4S
292.36
BGMSTNYJYPSLHN-UHFFFAOYSA-N
6-1-naphthalenylmethyl thio-9h-purine,6-naphthalen-1-ylmethylsulfanyl-7h-purine,6-naphthalen-1-ylmethyl sulfanyl-9h-purine,6-naphthylmethylthio purine,6-1-naphthylmethyl sulfanyl-9h-purine,6-naphthalen-1-ylmethyl thio-7h-purine,6-naphthalen-1-ylmethylsulfanyl-9h-purine
720937
6-(naphthalen-1-ylmethylsulfanyl)-7H-purine
C1=CC=C2C(=C1)C=CC=C2CSC3=NC=NC4=C3NC=N4

Negative allosteric modulator of 5-HT3

Negative allosteric modulator of 5-HT3 receptors (IC50 values are 0.36 and 0.73 μM in HEK293 cells transfected with human 5-HT3A and 5-HT3AB receptors respectively).
Quantity 10mg
Formula Weight 292.36
Percent Purity >97%
Chemical Name or Material PU 02

Recommended Storage : Store at +4°C