Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PU.1/Spi-1, Mouse anti-Human, Alexa Fluor 405, Clone: PU1/2146, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275766AF405
Description
PU.1/Spi-1 Monoclonal antibody specifically detects PU.1/Spi-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
PU.1/Spi-1 | |
PU1/2146 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SPI1 | |
Recombinant fragment (around aa 16-170) of human PU.1 protein (Exact sequence is proprietary) | |
Monoclonal | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
PU.1 is a member of the ETS family of transcription factors and is important for normal B-cell development. It is expressed in the myeloid lineage and in immature as well as mature B-lymphocytes, with the exception of plasma cells. PU.1 is expressed in germinal center B-cells and mantle B-cells. Various lymphomas are also positive for this marker. It is essential during early B-cell differentiation. The absence of PU.1 results in total block of B-cell development at the pre-pro stage. PU.1 plays a key role in normal myeloid differentiation, and regulates the expression of immunoglobulin and other genes that are important for B-cell development. |
Western Blot,Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
Purified,Conjugated | |
hematopoietic transcription factor PU.1, OF, PU.1, SFPI131 kDa transforming protein, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1 | |
Mouse | |
IgG2b Kappa | |
Primary | |
0.1 mL | |
Innate Immunity | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title