Learn More
Tocris™ PU 23
Multidrug resistance protein (MRP) 4 inhibitor
Manufacturer:  Tocris™ 6885/10
 View more versions of this product
Catalog No. 68-851-0
Add to cart
This item is not returnable. View return policy
Description
Description
PU 23 is a multidrug resistance protein (MRP) 4 inhibitor. Sensitizes MRP4-overexpressing cells to the anticancer agent 6-Mercaptopurine (Cat. No. 4103; 6-MP) (IC50 values are 14.11 and 6.06 μM in the absence and presence of compound, respectively). Increases accumulation of 6-MP in MRP4-overexpressing cells.
Specifications
Specifications
425.52
Multidrug resistance protein (MRP) 4 inhibitor
10 mg
C21H19N3O3S2
98%
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C