Learn More
BiolegendSupplier Diversity Partner Purified anti-α-Synuclein, aggregated DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 847801
Purified anti-α-Synuclein, aggregated [Syn-F1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: IHC-P, Dot Blot; Size: 25 µg
Catalog No. 50-114-5369