Learn More
Selleck Chemical LLCSupplier Diversity Partner Puerarin 10mg 3681-99-0 Kakonein  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S234610MG
Puerarin (Kakonein), an isoflavones found in the root of Radix puerariae, is a 5-HT2C receptor and benzodiazepine site antagonist.
Catalog No. 50-668-4