Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUF60 Mouse anti-Human, Mouse, Rat, DyLight 650, Clone: 917, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243764C

Catalog No. NBP243764C


Add to cart

Description

Description

PUF60 Monoclonal antibody specifically detects PUF60 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

PUF60
917
Western Blot
Purified
PUF60
Recombinant protein encompassing a sequence within the center region of human PUF60. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
FBP interacting repressor, FBP-interacting repressor, FIRpoly-U binding splicing factor PUF60, FLJ31379, FUSE-binding protein-interacting repressor, poly(U)-binding-splicing factor PUF60, poly-U binding splicing factor 60KDa, Ro ribonucleoprotein binding protein 1, Ro ribonucleoprotein-binding protein 1, Ro-binding protein 1, roBP1, ROBPI, siah binding protein 1,60 kDa poly(U)-binding-splicing factor, Siah-binding protein 1, siah-BP1, SIAHBP1pyrimidine tract binding splicing factor
Mouse
IgG2b
0.1 ml
Primary
22827
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit