Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCAR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550671
Description
The antibody detects endogenous levels of total PUMA gamma protein.
HCAR2 Polyclonal antibody specifically detects HCAR2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
HCAR2 | |
Polyclonal | |
Unconjugated | |
HCAR2 | |
G protein-coupled receptor 109A; G protein-coupled receptor 109B; G protein-coupled receptor HM74a; Gpr109; Gpr109a; Gpr109b; G-protein coupled receptor 109; G-protein coupled receptor 109A; G-protein coupled receptor HM74; G-protein coupled receptor HM74A; HCA2; Hcar2; HM74; HM74a; HM74b; hydroxycarboxylic acid receptor 2; hydroxy-carboxylic acid receptor 2; interferon-gamma inducible gene, Puma-g; mHM74b; Niacin receptor 1; Niacr1; Nicotinic acid receptor; Protein PUMA-G; PUMAG; PUMA-G; PUMA-G. GPR109; putative seven transmembrane spanning receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
338442, 369160, 80885 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q80Z39, Q8TDS4, Q9EP66 | |
HCAR2 | |
Synthetic peptide corresponding to a region derived from internal residues of human hydroxycarboxylic acid receptor 2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction