Learn More
Bellco Glass, Inc.Supplier Diversity Partner Pump Drive Only 1Rpm 115V  DFS Item
Manufacturer:  Bellco Glass, Inc.Supplier Diversity Partner 780711501
Must order with 7807-10000Torque: 224 in-oz, (16 kg-cm)Input: 115v ac ±10%, 60hzCurrent amps: .5Dimensions (lxwxh): 6" x 4" x 4.4"Weight: 2.3 kgs (5.1 lbs)Complete #: (pump head & pump drive), 7807-01115 with lighted on/off switch.6ft power cord.
Catalog No. 50-194-6373