Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURA Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18240020UL

 View more versions of this product

Catalog No. NBP18240020


Add to cart

Description

Description

PURA Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

PURA
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
PURA
Synthetic peptide towards Pura. Peptide sequence KPTYRNSITVPYKVWAKFGHTFCKYSEEMKKIQEKQREKRAACEQLHQQQ.
Immunogen affinity purified
RUO
Primary
5813
Western Blot
Western Blot 1:1000
NP_033015
PUR1PUR-ALPHA, PURALPHA, purine-rich element binding protein A, Purine-rich single-stranded DNA-binding protein alpha, transcriptional activator protein Pur-alpha
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit