Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters: PROMO

Product

Manufacturer:  GE Healthcare 67862504PM

Catalog No. 05-713-402LR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Diameter: 25mm
  • Sample volume up to 100mL
  • Low hold-up volumes for maximum sample recovery
  • Optional glass microfiber
  • Autoclave at 121ºC
Applications:
  • HPLC aqueous sample preparation
  • Biological sample preparation
  • Buffer solutions
  • Salt solutions
  • Tissue culture media
  • Irrigation solutions
  • Sterile isolation
Specifications

Specifications

Non-sterile
25mm
5.2 bar
Up to 100mL
4.2 sq. cm
Male Luer
Dimensions: 22.9 × 28.4mm
Syringe Filter
Depth Polypropylene (DpPP)
Pigment-free Polypropylene
0.45μm
Female Luer Lock
50
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit