Learn More
GE Healthcare Whatman™ Puradisc 25mm PTFE Syringe Filters: PROMO
Contains hydrophobic membrane that is chemically stable and inert. Whatman™ Puradisc 25mm PTFE Syringe Filters exhibit low protein binding, and its main application is filtration of non-aqueous samples.
Specifications
Diameter (Metric) 25mm
Material Polytetrafluoroethylene (PTFE)
Housing Material Pigment-free Polypropylene
Sterility Non-sterile
View More Specs
Products 2
Catalog Number Mfr. No. Pore Size Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size Quantity Price Quantity & Availability  
05-713-395LR View Documents GE Healthcare
67842501PM
0.1μm 50Pack This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
05-713-397LR View Documents GE Healthcare
67842504PM
0.45μm 50Pack This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • For filtering solvents, chemicals and nonaqueous solutions
  • Typical applications include HPLC, TLC, and GC sample clarification; HPLC solvent filtration
  • Made of hydrophobic polytetrafluoroethylene (PTFE)
  • Nonsterile, autoclavable
Specifications
Specifications
25mm
Polytetrafluoroethylene (PTFE)
Pigment-free Polypropylene
Non-sterile