Learn More
GE Healthcare Whatman™ Puradisc PES Syringe Filters: Nonsterile: PROMO
Designed for stability in low pH, have low levels of extractables, and exhibit low protein binding, making them suitable for many aqueous and organic solvents. Whatman™ Puradisc PES Syringe Filters: Nonsterile allow higher liquid flow than either PTFE or PVDF.
Manufacturer:  GE Healthcare 67942504PM
 View more versions of this product
Catalog No. 05-713-384LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Polyethersulfone (PES) membrane for filtering aqueous solutions
  • Low protein binding
  • Polypropylene housing
  • Female Luer Lock inlet
  • Male Luer outlet
Specifications
Specifications
Non-sterile
25mm
5.2bar
Pigment-free Polypropylene
0.45μm
Female Luer Lock
1000Pack
Syringe Filter
Polyethersulfone (PES)
For aqueous or solvent-based solutions, HPLC, TLC, GC sampling
Up to 100mL
4.2cm2
Male Luer