Learn More
GE Healthcare Whatman™ Puradisc PES Syringe Filters: Nonsterile: PROMO
Designed for stability in low pH, have low levels of extractables, and exhibit low protein binding, making them suitable for many aqueous and organic solvents. Whatman™ Puradisc PES Syringe Filters: Nonsterile allow higher liquid flow than either PTFE or PVDF.
Specifications
Diameter (Metric) 25mm
Material Polyethersulfone (PES)
Sterility Non-sterile
View More Specs
Products 2
Catalog Number Mfr. No. Pore Size Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size Quantity Price Quantity & Availability  
09-910-27LR View Documents GE Healthcare
67942502PM
0.2μm 1000Pack This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
05-713-384LR View Documents GE Healthcare
67942504PM
0.45μm 1000Pack This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Polyethersulfone (PES) membrane for filtering aqueous solutions
  • Low protein binding
  • Polypropylene housing
  • Female Luer Lock inlet
  • Male Luer outlet
Specifications
Specifications
25mm
Polyethersulfone (PES)
Non-sterile