Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.
Supplier: Cytiva 67882504
Description
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.
- Diameter: 25mm
- Sample volume up to 100mL
- Low hold-up volumes for maximum sample recovery
- Optional glass microfiber
- Autoclave at 121ºC
Applications:
- HPLC aqueous sample preparation
- Biological sample preparation
- Buffer solutions
- Salt solutions
- Tissue culture media
- Irrigation solutions
- Sterile isolation
Specifications
Puradisc 25 Depth Polypropylene Syringe Filter, 0.45 μm (200 pcs) | |
0.45 μm | |
4.2 cm2 | |
Female luer lock | |
Male luer | |
Syringe Filter | |
22.9 x 28.4 mm |
Depth Polypropylene (DpPP) | |
Non-sterile | |
Pigment Free Polypropylene (PP) | |
200 | |
5.2 bar (75 psi) | |
Up to 100 mL | |
25 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction