Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Puradisc 25mm PTFE Syringe Filters

Contains hydrophobic membrane that is chemically stable and inert. Whatman™ Puradisc 25mm PTFE Syringe Filters exhibit low protein binding, and its main application is filtration of non-aqueous samples.

Manufacturer:  GE Healthcare 67842501

 View more versions of this product

Catalog No. 05-713-395


Add to cart

Description

Description

  • For filtering solvents, chemicals and nonaqueous solutions
  • Typical applications include HPLC, TLC, and GC sample clarification; HPLC solvent filtration
  • Made of hydrophobic polytetrafluoroethylene (PTFE)
  • Nonsterile, autoclavable
Specifications

Specifications

Syringe Filter
25mm
5.2 bar (75 psi)
Pigment Free Polypropylene (PP)
Up to 100mL
4.2 cm2
Male luer
Up to 100 mL
50 Pieces
25 mm
Polytetrafluoroethylene (PTFE)
For HPLC, TLC, and GC sample clarification; HPLC solvent filtration
Whatman Puradisc syringe filters from GE Healthcare’s Life Sciences business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 mL.
0.1μm
Female luer lock
Puradisc 25 mm Polytetrafluoroethylene Syringe Filter, 0.1 μm, nonsterile (50 pcs)
22.9 x 28.4 mm
Non-sterile
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit