Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Puradisc 25mm PTFE Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Contains hydrophobic membrane that is chemically stable and inert. Whatman™ Puradisc 25mm PTFE Syringe Filters exhibit low protein binding, and its main application is filtration of non-aqueous samples.
$334.00 - $2350.00
Specifications
Diameter (Metric) 25mm
Material Polytetrafluoroethylene (PTFE)
Housing Material Pigment free polypropylene (PP)
Sterility Non-sterile
View More Specs
Products 10
Catalog Number Mfr. No. Pore Size Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size Quantity Price Quantity & Availability  
05-713-851 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
67982501
0.1μm 1000 Pieces Pack of 1000 for $2,350.00
Add to cart
05-713-392 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
67982504
0.45μm 1000 Pieces Pack of 1000 for $2,350.00
Add to cart
05-713-395
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
67842501
0.1μm 50 Pieces Pack of 50 for $334.00
Add to cart
05-713-396
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
67842502
0.2μm 50 Pieces Pack of 50 for $334.00
Add to cart
05-713-397 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
67842504
0.45μm 50 Pieces Pack of 50 for $334.00
Add to cart
05-713-398
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
67842510
1.0μm 50 Pieces Pack of 50 for $334.00
Add to cart
05-713-399
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
67852502
0.2μm 200 Pieces Pack of 200 for $876.00
Add to cart
22-022733 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
67982502
0.2μm 1000 Pieces Case of 1000 for $2,350.00
Add to cart
09302216
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
67982510
1.0μm 1000 Pieces Pack of 1000 for $2,350.00
Add to cart
05-713-400 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
67852504
0.45μm 200 Pieces Pack of 200 for $876.00
Add to cart
Description
Description
Contains hydrophobic membrane that is chemically stable and inert. Whatman™ Puradisc 25mm PTFE Syringe Filters exhibit low protein binding, and its main application is filtration of non-aqueous samples.
  • For filtering solvents, chemicals and nonaqueous solutions
  • Typical applications include HPLC, TLC, and GC sample clarification; HPLC solvent filtration
  • Made of hydrophobic polytetrafluoroethylene (PTFE)
  • Nonsterile, autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
25mm
Polytetrafluoroethylene (PTFE)
Pigment free polypropylene (PP)
Non-sterile