Learn More
Whatman™ Puradisc Syringe Filters, Nonsterile: PROMO OFFER
Low hold-up volume—less than 0.1mL with air purge
Manufacturer:  Whatman™ 67852504PM
 View more versions of this product
Catalog No. 05-713-400PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Syringe Filter
Polytetrafluoroethylene (PTFE)
Polypropylene
female Luer-Lok
200/pk.
25mm
For HPLC, TLC, and GC sample clarification; HPLC solvent filtration
0.45μm
male Luer slip