Learn More
Biolegend Purified anti-human/mouse SSEA-3
Supplier:  Biolegend 330302
Encompass_Preferred
Purified anti-human/mouse SSEA-3 [MC-631]; Isotype: Rat IgM, κ; Reactivity: Human, African Green, Mouse; Apps: FC, ICC, ELISA; Size: 100 ug
Catalog No. 50-167-557
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.