Learn More
BUCHI Pure Chromatography System Available on GSA/VA Contract for Federal Government customers only.
BUCHI Pure Chromatography System is extremely compact and can be used outside of the fume hood. Choose from flash systems or combination flash/prep HPLC capable of small to large scale purification of complex mixtures. UV detection is standard or choose to add ELS detection to detect all compounds.
Manufacturer:  BUCHI 11C83500
 View more versions of this product
Catalog No. 10-987-139
Add to cart
Description
Description
  • Pure space. Pure safety. Pure simplicity.
  • Small footprint onboard solvent platform
  • Cartridge/ column holders on front of the system
  • Lowest sample loss for ELSD (30 µL/min)
  • Closed fraction collector bay
  • Leak and pressure sensors for safety
  • Remote control capable from PC, tablet or smartphone
  • All important parts are easily accessable for cleaning
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
    Specifications
    Specifications
    680 mm
    100 to 240 V ± 10 %
    33 kg
    Prep HPLC/UV-Vis/ELSD
    0 to 300 bar (0 to 4350 psi)
    570 mm
    365 mm
    Prep HPLC
    300 bar (4350 psi)