Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Pure Chromatography Collection Rack

Pure chromatography collection rack

$379.50

Specifications

For Use With (Application) Purification
View More Specs

Products 4
Catalog Number Mfr. No. Size (Metric) Tube Rack Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Tube Rack Price Quantity & Availability  
05-405-479
View Documents
BUCHI
11066674
16 x 125 mm
Each for $379.50
Add to cart
 
05-405-480
View Documents
BUCHI
11066675
16 x 150 mm
Each for $379.50
Add to cart
 
05-405-481
View Documents
BUCHI
11066676
18 x 150 mm
Each for $379.50
Add to cart
 
05-405-482
View Documents
BUCHI
11066677
25 x 150 mm
Each for $379.50
Add to cart
 
Description

Description

  • Pure chromatography system rack
  • For glass tubes or squared bottles
Specifications

Specifications

Purification
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit