Learn More
Saint-Gobain Pure-Fit™ SC Sterile Connectors
Create true sterile connections anywhere – even outside the clean room environment. Saint-Gobain Pure-Fit™ SC secure aseptic connections incorporate Pure-Fit™ SIB™ technology, which provides a seamless transition between tubings and fittings to maintain complete fluid integrity.
Manufacturer:  Saint-Gobain PFSC1.00IHM
 View more versions of this product
Catalog No. 13-417-237
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The silicone valve design at the heart of Pure-Fit™ SC allows for easy assembly and leaves no gaps or residue, eliminating problems associated with other systems. The locking mechanism virtually eliminates the possibility of operator error, and also simplifies validation.
  • Laser etched for complete lot traceability
  • Triple-redundant locking mechanism simplifies validation, reducing chances of operator error
  • Patented Pure-Fit™ SIB™ (smooth inner bore) technology on barb connections ensures a completely smooth fluid path
  • Available in interchangeable 3/4 in. and 1 in. sizes for greater flow volume
  • Available in three interchangeable sizes (1/4 in., 3/8 in., 1/2 in.) for step-down requirements
  • All components made of fully characterized, animal derivative-free and BPA-free materials
  • Easy-to-see laser-etched visual indicators confirm proper engagement
  • Can be sterilized via gamma irradiation or autoclaving
Specifications
Specifications
USP Class VI, ISO 10993, EP 3.1.9
Autoclavable
5.79 in.
20 psi (1.38 bar)
40°C
0°C
2.00 in. (5.1cm)
Red/Clear
PVDF Housing, C-Flex Caps
Inner Housing
104°F
32°F
1 in. (25.4mm) Hose Barb