Learn More
Saint-Gobain Pure-Fit™ SIB™ 360° Jaw Set
Table top 360° jaw sets for easy assembly, removal and handling of Pure-Fit SIB hose barb connectors. Jaw sets are available for 1/16 in., 1/8 in., 3/16 in. and 5/16 in. ID tubing. Can also be used with BarbLock™ ultra-secure tubing retainers.
Manufacturer:  Saint-Gobain PFT062JAW360
 View more versions of this product
Catalog No. 13-417-336
Add to cart
Description
Description
Pure-Fit SIB is a smooth inner bore (SIB) hose-barb connector that provides a seamless transition between tubing and fittings throughout the fluid path. Pure-Fit SIB eliminates entrapment and leak points that can occur with traditional assembly systems. The unique internal design eliminates voids or dead space that can lead to turbulence or stagnation of fluid flow creating the potential for bacterial growth.
  • 062 Series = 1/16 in. ID tubing
  • 125 Series = 1/8 in. ID tubing
  • 187 Series = 3/16 in. ID tubing
  • 312 Series = 5/16 in. ID tubing
Specifications
Specifications
Pure-Fit SIB Connectors, BarbLock Retainers and Saint-Gobain Sani-Tech, C-Flex and PharMed Flexible Tubing
1/16 in. Tubing
062 Series
360° Jaw Set
Assembly and Removal of Tubing Connectors and Retainers