Learn More
Saint-Gobain Pure-Fit™ SIB™ Reducer Series
Utilizes the patented BarbLock™ retention and seal system. Pure-Fit™ SIB™ provides a seamless transition that maintains fluid integrity and leak-free connections with complete biocompatibility for the most demanding applications and stringent test protocols.
$273.50 - $530.00
Specifications
Product Line Reducer Series
Certifications/Compliance USP Class VI, ISO
For Use With Saint-Gobain Sani-Tech™, C-Flex™ and PharMed™ flexible tubing
Maximum Pressure 250 psi
View More Specs
Products 14
Catalog Number Mfr. No. Diameter (English) Inner Material Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Material Price Quantity & Availability  
13-417-240
View Documents
Saint-Gobain
PFR062X250PPAF
1/16 x 1/4 in PP
Pack of 25 for $273.50
Add to cart
13-417-250
View Documents
Saint-Gobain
PFR187X312PPAF
3/16 x 5/16 in. PP
Pack of 25 for $310.50
Add to cart
13-417-285
View Documents
Saint-Gobain
PFR062X125PVDF
1/16 x 1/8 in PVDF
Pack of 25 for $467.50
Add to cart
13-417-286
View Documents
Saint-Gobain
PFR062X250PVDF
1/16 x 1/4 in PVDF
Pack of 25 for $467.50
Add to cart
13-417-292
View Documents
Saint-Gobain
PFR125X187PVDF
1/8 x 3/16 in PVDF
Pack of 25 for $487.00
Add to cart
13-417-293
View Documents
Saint-Gobain
PFR125X250PVDF
1/8 x 1/4 in. PVDF
Pack of 25 for $487.00
Add to cart
13-417-297
View Documents
Saint-Gobain
PFR187X312PVDF
3/16 x 5/16 in PVDF
Pack of 25 for $489.00
Add to cart
13-417-298
View Documents
Saint-Gobain
PFR187X375PVDF
3/16 x 3/8 in PVDF
Pack of 25 for $489.00
Add to cart
13-417-318
View Documents
Saint-Gobain
PFR375X500PVDF
3/8 x 1/2 in PVDF
Pack of 25 for $527.50
Add to cart
13-417-327
View Documents
Saint-Gobain
PFR250X500PVDF
1/4 x 1/2 in PVDF
Pack of 25 for $530.00
Add to cart
13-417-259
View Documents
Saint-Gobain
PFR250X375PPAF
1/4 x 3/8 in PP This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
13-417-271
View Documents
Saint-Gobain
PFR375X500PPAF
3/8 x 1/2 in PP This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
13-417-280
View Documents
Saint-Gobain
PFR250X500PPAF
1/4 x 1/2 in PP This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
13-417-245
View Documents
Saint-Gobain
PFR125X250PPAF
1/8 x 1/4 in PP This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Pure-Fit SIB eliminates entrapment and leak points that can occur with traditional assembly systems. The unique internal design eliminates voids or dead space that can lead to turbulence or stagnation of fluid flow creating the potential for bacterial growth.
  • Available in both animal-free polypropylene (PP) and polyvinylidene fluoride (PVDF)
    • All materials completely animal-free and BPA-free
    • PP is gamma stable, PVDF is autoclavable and gamma stable
  • Fully compatible with Pure-Fit and BarbLock retainers for complete 360° compression
  • USP Class VI and ISO compliant; Manufactured and packaged in a certified clean room
  • Available in a variety of sizes
Specifications
Specifications
Reducer Series
USP Class VI, ISO
Saint-Gobain Sani-Tech™, C-Flex™ and PharMed™ flexible tubing
250 psi