Learn More
Saint-Gobain Pure-Fit™ TC Tube Clamps, Polypropylene
Pure-Fit™ TC tube clamps for the pharmaceutical industry have an innovative design that allows for complete flow stoppage. The clamp can be installed over the tube in the normal fashion or put together after the tube is already part of a completed assembly. 
$254.50 - $2300.00
Specifications
Autoclavable Autoclavable
Material Polypropylene
Size Standard
Temperature (English) Resistant 200°F
Temperature (Metric) Resistant 93°C
View More Specs
Products 9
Catalog Number Mfr. No. Color Type For Use With (Equipment) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Color Type For Use With (Equipment) Quantity Price Quantity & Availability  
13-417-341
View Documents
Saint-Gobain
PFTC375250
White Assembled Fits tubing 1/8 to 3/8 in. OD 250
Pack of 250 for $1,910.00
Add to cart
13-417-342
View Documents
Saint-Gobain
PFTC375UA
White Unassembled Fits tubing 1/8 to 3/8 in. OD 250
Pack of 250 for $1,849.00
Add to cart
13-417-345
View Documents
Saint-Gobain
PFTC750
White Assembled Fits tubing 3/8 to 3/4 in. OD 25
Pack of 25 for $254.50
Add to cart
13-417-346
View Documents
Saint-Gobain
PFTC750R RED
Red Assembled Fits tubing 3/8 to 3/4 in. OD 25
Pack of 10 for $372.00
Add to cart
13-417-347
View Documents
Saint-Gobain
PFTC750G GREEN
Green Assembled Fits tubing 3/8 to 3/4 in. OD 25
Pack of 10 for $369.00
Add to cart
13-417-348
View Documents
Saint-Gobain
PFTC750BU BLUE
Blue Assembled Fits tubing 3/8 to 3/4 in. OD 25
Pack of 10 for $369.00
Add to cart
13-417-349
View Documents
Saint-Gobain
PFTC750250
White Assembled Fits tubing 3/8 to 3/4 in. OD 250
Pack of 250 for $2,300.00
Add to cart
13-417-350
View Documents
Saint-Gobain
PFTC750PPUA
White Unassembled Fits tubing 3/8 to 3/4 in. OD 250
Pack of 250 for $2,230.00
Add to cart
13-417-340
View Documents
Saint-Gobain
PFTC375
White Assembled Fits tubing 1/8 to 3/8 in. OD 25 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Pure-Fit™ TC tube clamps feature an outer body with no sharp edges, eliminating the potential for snags, scratches or punctures; a press-down locking system that is fully encapsulated to guard against untimely or unwanted release; a side release mechanism that facilitates secure, single-handed installation and operation; and an integral bore hole for absolute lock-out capabilities.
  • Provides complete fluid stoppage
  • Smooth edges and corners help prevent punctures and ruptures
  • Can be installed on or over tubing or finished assemblies
  • High degree of leverage accommodates tubing with different durometers
  • Single-hand installation capability for ease of operation
  • Unique top locking mechanism and side release prevent unwanted openings
  • Constructed from high performance thermoplastic material that is completely animal derivative- and BPA-free
  • Fully autoclavable and sterilizable, and meets all USP Class VI criteria
Specifications
Specifications
Autoclavable
Standard
93°C
Polypropylene
200°F