Learn More
BiolegendSupplier Diversity Partner Purified Mouse IgG2b, κ Isotype Ctrl DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 400301
Purified Mouse IgG2b, κ Isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: FC, ICFC, ICC, IF, IHC, IP, WB; Size: 50 ug
Catalog No. NC0138234