Learn More
MilliporeSigma™ Supelco™ Purge/Trap D
Specific configurations available for a variety of instruments
$45.50 - $103.00
Specifications
For Use With (Application) Industrial Hygiene
Type Purge/Trap D
View More Specs
Products 3
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
11-101-0588
View Documents
MilliporeSigma™ Supelco™
28981U
Tekmar™ Stratum Each for $103.00
Add to cart
11-101-1473
View Documents
MilliporeSigma™ Supelco™
21062U
Tekmar™ Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000 (Straight, 12 in., Swagelok Fitting), HT-3 Each for $45.50
Add to cart
11-101-1368
View Documents
MilliporeSigma™ Supelco™
24913U
Tekmar™ 3000, 3100 (Straight, 12 in., Valco Fitting) Each for $51.20
Add to cart
Description
Description
  • Tubes used in purge and trap analyses generally are packed with multiple beds of adsorbent materials
  • A single tube can trap a broad range of polar and nonpolar, high and low molecular weight compounds
  • Each bed protects the next, increasingly active bed, by preventing compounds from being held so strongly that they cannot be desorbed quickly without decomposition
  • During the purge phase of sampling, lower molecular weight compounds pass through the initial adsorbent beds but are trapped by succeeding beds
  • During desorption, the carrier gas passes through the trap in the reverse direction of purge flow, so that higher molecular weight compounds never come in contact with the stronger (innermost) adsorbents
Specifications
Specifications
Industrial Hygiene
Purge/Trap D