Learn More
AccuStandard, IncSupplier Diversity Partner PURGEABLES EPA 624 1X1ML DFS Item
Manufacturer:  AccuStandard, IncSupplier Diversity Partner M624
Purgeables - EPA Method 624. 5 x 1 mL. 0.2 mg/mL in MeOH.
Catalog No. NC0101580