Learn More
BiolegendSupplier Diversity Partner PURIFIED ANTI-HUMAN KLRG1 MAFA  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 368602
Purified anti-human KLRG1 (MAFA) Antibody, 100ug.Killer cell lectin-like receptor subfamily G member (KLRG1) is a 30 kD, type II membrane glycoprotein with one C-type lectin domain and one immunoreceptor tyrosine-based inhibitory motif (ITIM). KLRG1 is expressed by subsets of natural killer cells (NKs) and effector and memory T cells. It inhibits cell activation and proliferation and is also a marker of T cell senescence. The binding of KLRG1 to E-, N-, or R- cadherins blocks phosphorylation of AKT and increases the expression of cell cycle inhibitors.
Catalog No. NC1480766