Learn More
BiolegendSupplier Diversity Partner Purified anti-STAT5  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 660202
Purified anti-STAT5 [9C8B50]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse; Apps: WB, ICC, IP; Size: 100 ug
Catalog No. 50-712-611