Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Purified CD40LG (AAH74950.1, 116 a.a. - 261 a.a.) human Recombinant Protein with His-Flag-StrepII tag at N-terminus expressed in human cells

Used for ELISA, PI, SDS-PAGE, WB

Manufacturer:  Abnova™ H00000959H01

Catalog No. 89-938-708


Add to cart

Description

Description

The protein encoded by this gene is expressed on the surface of T cells. It regulates B cell function by engaging CD40 on the B cell surface. A defect in this gene results in an inability to undergo immunoglobulin class switch and is associated with hyper-IgM syndrome. [provided by RefSeq]

Sequence: GDQNPQIAAHVISEASSKTTSVLQWAEKGYYTMSNNLVTLENGKQLTVKRQGLYYIYAQVTFCSNREASSQAPFIASLCLKSPGRFERILLRAANTHSSAKPCGQQSIHLGGVFELQPGASVFVNVTDPSQVSHGTGFTSFGLLKL
Specifications

Specifications

Transfection of pSuper-CD40LG plasmid into HEK293T cell, and the expressed protein was purified by Strep-Tactin affinity column.
≥ 10 ug/ml
Liquid
959
CD40LG (Human) Recombinant Protein
Strep-Tactin affinity columns
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD154/CD40L/HIGM1/IGM/IMD3/T-BAM/TNFSF5/TRAP/gp39/hCD40L
CD40LG
Human
His-Flag-StrepII
AAH74950.1
Enzyme-linked Immunoabsorbent Assay, Protein Interaction, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis, Western Blot
Liquid
21.34kDa
Transfection of pSuper-CD40LG plasmid into HEK293T cell, and the expressed protein was purified by Strep-Tactin affinity column.
SDS-PAGE and Western Blot
GDQNPQIAAHVISEASSKTTSVLQWAEKGYYTMSNNLVTLENGKQLTVKRQGLYYIYAQVTFCSNREASSQAPFIASLCLKSPGRFERILLRAANTHSSAKPCGQQSIHLGGVFELQPGASVFVNVTDPSQVSHGTGFTSFGLLKL
RUO
CD40LG
Not Tested
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit