Learn More
ZeptoMetrix Purified Viral Lysate, Human Immunodeficiency Virus Type 1 (HIV-1)
Can be utilized as antigen, source for purification of viral proteins or for detection of viral antibodies
Supplier:  ZeptoMetrix 0810027
 View more versions of this product
Catalog No. 22-157-411
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Available in 1mg vials
Specifications
Specifications
2
0.5 to 3mg/mL
Viral Antibodies
Vial
Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay
1 mg
ISO, 13485
-65°C
Solution
Purified Viral Lysate
Sucrose density gradient ultracentrifugation
Mn
Safety and Handling
Safety and Handling
ShelfLife : 1 Year