Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ Purifier™ Horizontal Clean Bench, 6ft.

When using a Labconco Purifier™ Horizontal Clean Bench, workers are protected from particulate contamination.
Supplier: Labconco™ 3873021
Alert:
Units do not protect user and should not be used in conjunction with biohazardous material, toxins or radionuclides.
Requires : Supporting base (sold separately).
Description
- Horizontal laminar flow
- Double-gasket, negative pressure HEPA filter leak protection
- ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
- Disposable prefilters (2 each on 3 ft. and 4 ft. models; 3 each on 6 ft. models; 4 each on 8 ft. models)
- Variable speed built-in motor/blower(s) with vibration isolation pads
- Differential pressure gauge
- Type 304 brushed stainless-steel work surface
- Tempered safety glass side panels with utility ports with iris openings
- UV-resistant, dry powder epoxy-coated steel exterior
- Front-mounted light and blower switches
- W x H (Exterior): 75 x 48.4 in.
Order Info
Shipped assembled.
Specifications
Fluorescent | |
66 cm | |
122.9 cm | |
Domestic | |
299 kg | |
190.5 cm |
26 in. | |
48.4 in. | |
60 Hz | |
660 lb. | |
75 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title