Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Horizontal Clean Bench, 6ft.
SDP

When using a Labconco Purifier™ Horizontal Clean Bench, workers are protected from particulate contamination.

$18101.00 - $22239.00

Specifications

Width (English) Exterior 75 in.
Width (Metric) Exterior 190.5cm
Height (English) Exterior 48.4 in.
Height (Metric) Exterior 122.9cm
Voltage 115V
View More Specs

Alert:

Units do not protect user and should not be used in conjunction with biohazardous material, toxins or radionuclides.

Requires: Supporting base (sold separately).


Products 2
Catalog Number Mfr. No. Depth (Metric) Interior Electrical Requirements Plug Type Price Quantity & Availability  
Catalog Number Mfr. No. Depth (Metric) Interior Electrical Requirements Plug Type Price Quantity & Availability  
13-118-13-1
SDP
View Documents
Labconco™
3873021
66cm 115V, 60Hz, 6A Domestic
Each for $22,239.00
Add to cart
 
13-118-13-0
SDP
View Documents
Labconco™
3873020
53cm 115V, 60Hz, 6A Domestic
Each for $18,101.00
Add to cart
 
Description

Description

  • Horizontal laminar flow
  • Double-gasket, negative pressure HEPA filter leak protection
  • ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
  • Disposable prefilters (2 each on 3 ft. and 4 ft. models; 3 each on 6 ft. models; 4 each on 8 ft. models)
  • Variable speed built-in motor/blower(s) with vibration isolation pads
  • Differential pressure gauge
  • Type 304 brushed stainless-steel work surface
  • Tempered safety glass side panels with utility ports with iris openings
  • UV-resistant, dry powder epoxy-coated steel exterior
  • Front-mounted light and blower switches
  • W x H (Exterior):  75 x 48.4 in.

Order Info

Shipped assembled.

Specifications

Specifications

75 in.
48.4 in.
115V
99.99%
CAN/CSA C22.2, ETL, UL, CE
Fluorescent
Type 304 Brushed Stainless Steel
190.5cm
122.9cm
Utility
15A
115V, 60Hz, 6A
HEPA
Domestic
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit