Puritan™ ESK™ Sampling Kits with Pre-filled Letheen Broth

For environmental surface sampling with Letheen Broth

Manufacturer: Puritan™ 2583010 PDB LB

 View more versions of this product

Catalog No. 14-960-220

  Check Availability
Add to Cart

    Specifications

    Diameter (Metric) Handle 2.54mm
    Diameter (English) Tip 0.2 in.
    Diameter (Metric) Tip 5.1mm
    Certifications/Compliance ISO 9001:2008, ISO 13485:2003
    Capacity (English) 0.33 oz.
    Material Plastic
    Type Sampling Kit
    Length (Metric) 103cm
    Sterile Yes
    Includes Prefilled letheen broth
    Capacity (Metric) 10mL
    Length (English) 4 in.
    Length (English) Handle 3.125 in.
    Length (English) Tip 0.68 in.
    Length (Metric) Handle 79.3mm
    Length (Metric) Tip 1.73cm
    For Use With (Application) Environmental surface sampling with letheen broth and detection of microorganisms
    Capacity 0.33 oz., 10mL
    Shelf Life 18 Months
    Tip Material Polyester
    Quantity 300 Case
    Diameter (English) Handle 0.1 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit