Learn More
Puritan™ UniTranz-RT Transport Systems
For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Manufacturer:  Puritan™ UT362
 View more versions of this product
Catalog No. 22-027-107
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Hank's Balanced Salts, Bovine Serum Albumin, Gelatin, Sucrose, L-Cysteine, L-Glutamic Acid, Colistin, Amphotericin B, Vancomycin, Sodium Bicarbonate and HEPES Sodium Salt
Individually-wrapped
Two regular Polyester tipped
CE
Polystyrene
7.3 ±0.2
Polyester
UniTranz-RT Transport System
300 Case
2 to 25°C
Collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Two regular Polyester tip swabs, wrapped
Yes
Safety and Handling
ShelfLife : 18 Month(s)