Puritan™ UniTranz-RT Transport Systems

For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma

Manufacturer: Puritan™ UT366

 View more versions of this product

Catalog No. 22-027-109

  Check Availability
Add to Cart

    Specifications

    Product Type UniTranz-RT Transport System
    Certifications/Compliance CE
    For Use With (Application) Collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
    Includes One regular Polyester tip swab and one minitip swab, wrapped
    pH 7.3 ±0.2
    Sterile Yes
    Tip Material Polyester
    Packaging Individually-wrapped
    Media Type Hank's Balanced Salts, Bovine Serum Albumin, Gelatin, Sucrose, L-Cysteine, L-Glutamic Acid, Colistin, Amphotericin B, Vancomycin, Sodium Bicarbonate and HEPES Sodium Salt
    Shelf Life 18 Month(s)
    Handle Material Aluminum/Plastic
    Quantity 300 Case
    Swab Configuration One minitip and one regular Polyester tipped
    Storage Requirements 2 to 25°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit