Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Puritan MRSA Transport Medium

For the isolation of specimens containing Staphylococcus aureus spp, particularly methicillin-resistant Staphylococcus aureus (MRSA) as well as in vitro diagnostic use.

$68.80 - $76.00

Specifications

Format Tube, Tubes
Cap Type Screw Cap
Color Clear Tube
Sterility Sterile
Storage Requirements 2-25°C
View More Specs

Products 2
Catalog Number Mfr. No. Fill Volume Price Quantity & Availability  
Catalog Number Mfr. No. Fill Volume Price Quantity & Availability  
22-029-647 View Documents Puritan™
MRSA200
2mL Case of 50 for $68.80
Add to cart
 
22-029-648 View Documents Puritan™
MRSA500
5mL Case of 50 for $76.00
Add to cart
 
Specifications

Specifications

Tube, Tubes
Clear Tube
2-25°C
Screw Cap
Sterile
USA
Safety and Handling

Safety and Handling

ShelfLife : 18 months from date of manufacture

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit