Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ PurFlock Ultra™ and HydraFlock™ Flocked Swabs with Elongated Tips - Non-Sterile

Polystyrene handle available with breakpoint (at 30, 80 or 100mm)

Manufacturer:  Puritan™ 3806H

 View more versions of this product

Catalog No. 22-029-521


Add to cart

Description

Description

A new era in flock swab technology. HydraFlock provides superior collection and release of specimens. The unique microstructure of Hydra’s multi-length fibers are designed to enhance rapid absorption and quick release of biological specimens.; For buccal cell collection; Specimen collection; Tip material is comprised of polyester bicomponent fiber; Made in USA

Compliance

FDA Good Manufcturing Practices (GMP)

Specifications

Specifications

CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
Collection of Microorganisms, Buccal cells and forensic evidence
152.4mm
80 or 100mm
17.39mm
0.219 in.
Non-sterile
Elongated flocked
6 in.
HydraFlock™ Flocked Swab
3.14 or 4 in.
0.685 in.
5.5mm
Bag
Polystyrene
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit