Learn More
Puritan™ Puritan Regan Lowe Medium
For the transport and selective isolation of Bordetella pertussis and Bordetella parapertussis species as well as in vitro diagnostic use.
Manufacturer:  Puritan™ RL200
 View more versions of this product
Catalog No. 22-029-651
Add to cart
This item is not returnable. View return policy
Description
Description
2ml Regan-Lowe semi-solid transport medium.  12x80mm screw-cap polypropylene tube, white cap.  Semi-solid enrichment medium intended for transporting specimens containing Bordetellaspecies, the causative agent for whooping cough.  For In Vitro Diagnostic Use.  Premium medical grade plastic components to improve product shelf life.  Made in USA
Specifications
Specifications
Regan Lowe Semi-Solid Transport Medium
White
Regan Lowe Semi-Solid Transport Medium
Incubate aerobically at 35°C - 36°C. To avoid drying, agar plastes should be placed in a moist chamber with a damp filter paper.
US
Yes
2 mL
Tube
Sterile
Screw
Clear tube
For at least 7 days
80 mm
50
12 x 80 mm
50 Bulk
Regan Lowe Semi-Solid Transport Medium
Safety and Handling
Safety and Handling
ShelfLife : 18 Months from DOM