Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puromycin aminonucleoside, 98%, Alfa Aesar™

Click to view available options
Quantity:
1g
58-60-6
C12H18N6O3
294.315
MFCD00063462
RYSMHWILUNYBFW-GRIPGOBMSA-N
3?-Amino-3?-deoxy-N^6,N^6-dimethyladenosine
6020
CHEBI:42839
(2R,3R,4S,5S)-4-amino-2-[6-(dimethylamino)purin-9-yl]-5-(hydroxymethyl)oxolan-3-ol
CN(C)C1=NC=NC2=C1N=CN2C3C(C(C(O3)CO)N)O
58-60-6
C12H18N6O3
294.315
MFCD00063462
RYSMHWILUNYBFW-GRIPGOBMSA-N
3?-Amino-3?-deoxy-N^6,N^6-dimethyladenosine
6020
CHEBI:42839
(2R,3R,4S,5S)-4-amino-2-[6-(dimethylamino)purin-9-yl]-5-(hydroxymethyl)oxolan-3-ol
CN(C)C1=NC=NC2=C1N=CN2C3C(C(C(O3)CO)N)O

Used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.

Applications
Puromycin aminonucleoside is used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.

Solubility
Soluble in water at 50mg/ml. May require heating

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Beilstein 93902
Solubility Information Soluble in water at 50mg/mL. May require heating
Formula Weight 294.31
Physical Form Powder
Percent Purity 98%
Color White
Melting Point 215°C to 216°C
Quantity 1g
Flash Point >110°C(230°F)
Chemical Name or Material Puromycin aminonucleoside

GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AU7337000

TSCA : TSCA

storageNote1 : 2° to 8°C