Puromycin Dihydrochloride, Fisher BioReagents

Inhibits protein synthesis

$282.32

Chemical Identifiers

Chemical Name or Material Puromycin Dihydrochloride
CAS [58-58-2]
Physical Form Dry, White or off-white powder
Assay Percent Range > 98 %
Molecular Formula C22H29N7O5·2HCl
Formula Weight 544.44
Merck Index 15, 8054
View More Specs

  • Products 1
  • Description & Specifications
  • Safety and Handling
Products
Catalog Number Description Price Quantity Availability  

BP2956-100

SDS
Clear Glass; 100mg
Each for $282.32
Check Availability
Add to Cart
Descriptions & Specifications
Safety and Handling

Related Products