Learn More
MilliporeSigma™ Purospher™ STAR Phenyl (5µm) LiChroCART™ HPLC Cartridge
Provides the required pH stability for 99% of common applications. MilliporeSigma Purospher™ STAR Phenyl (5µm) LiChroCART™ HPLC Cartridge provides the power to separate the most difficult compound mixtures with optimal, reproducible results.
$654.65 - $731.69
Specifications
Column Type LiChroCART™ HPLC cartridge
Technique RP (reversed-phase)
Stationary Phase Phenyl
Particle Size 5 µm
View More Specs
Products 2
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
M1519210001 View Documents MilliporeSigma™
1.51921.0001
250 mm Each for $731.69
Add to cart
M1519220001 View Documents MilliporeSigma™
1.51922.0001
150 mm Each for $654.65
Add to cart
Specifications
Specifications
LiChroCART™ HPLC cartridge
RP (reversed-phase)
Phenyl
5 µm