Learn More
MilliporeSigma™ Purospher™ STAR RP-18 (5µm) Endcapped Hibar™ RT HPLC Column
Provides the required pH stability for 99% of common applications. MilliporeSigma Purospher™ STAR RP-18 (5µm) Endcapped Hibar™ RT HPLC Column provides the power to separate the most difficult compound mixtures with optimal, reproducible resu
$711.73 - $2450.88
Specifications
Column Type Hibar™ HPLC column
Technique RP (reversed-phase)
Stationary Phase RP18
Particle Size 5 µm
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity & Availability  
M1519140001 View Documents MilliporeSigma™
1.51914.0001
4.6 mm 125 mm Each for $711.73
Add to cart
M1519150001 View Documents MilliporeSigma™
1.51915.0001
10 mm 250 mm Each for $2,450.88
Add to cart
Specifications
Specifications
Hibar™ HPLC column
RP (reversed-phase)
RP18
5 µm