Learn More
MilliporeSigma™ Purospher™ STAR RP-8 (2µm) Hibar™ HR UHPLC Column
Provides the required pH stability for 99% of common applications. MilliporeSigma Purospher™ STAR RP-8 (2µm) Hibar™ HR UHPLC Column provides the power to separate the most difficult compound mixtures with optimal, reproducible results. Avail
$619.61 - $726.66
Specifications
Column Type Hibar™ UHPLC column
Technique RP (reversed-phase)
Stationary Phase RP8
Particle Size 2 µm
View More Specs
Products 2
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
M1506290001 View Documents MilliporeSigma™
1.50629.0001
100 mm Each for $726.66
Add to cart
M1506300001 View Documents MilliporeSigma™
1.50630.0001
50 mm Each for $619.61
Add to cart
Specifications
Specifications
Hibar™ UHPLC column
RP (reversed-phase)
RP8
2 µm