Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Purple Caps

Use with empty FastPrep™ tubes. MP Biomedicals Purple Caps are screw scaps with an O-ring for use with 2mL microfuge tubes. These caps are sold separately for those who wish to fill their own Lysing Matrix E tubes for prepare samples for homogenizat

$263.00 - $1391.00

Specifications

Storage Requirements Store at Room Temperature (15–30°C).
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP115064005 View Documents MP Biomedicals™
115064005
1000 Caps Each for $263.00
Add to cart
 
MP115064010 View Documents MP Biomedicals™
115064010
5000 Caps Each for $1,391.00
Add to cart
 
Description

Description

Purple Caps fit empty FastPrep™ tubes and are used in FastPrep systems. Add sample to empty tube, add Lysing Matrix E material and lysing buffer, and apply cap. Process using FastPrep instrument or other bead-beater to lsye, grind or homogenize sample. Purple caps are also found in the Fast DNA™ Spin Kit for Soil and Fast RNA™ Pro Soil Kit.

Specifications

Specifications

Store at Room Temperature (15–30°C).
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit