Purus™ PureTouch Nylon Glove Liners
Comfortable barrier reduces perspiration, wicks moisture and prevents skin irritation while limiting wear and tear on gloves
Manufacturer: Purus International Inc GLFFM
 View more versions of this product
Catalog No. 19-125-916
  Check Availability
Add to Cart
    Specifications
    Product Type Full glove liner
    Certifications/Compliance ISO 9002, ASTM AQL 1.5
    Disposable No
    For Use With (Application) Prevents skin irritation without diminishing tactile sensitivity, reduces perspiration and wicks moisture during extended use
    For Use With (Equipment) For use under Latex, Nitrile, PVC, and all exam gloves
    Material Nylon
    Size Medium
    Surface Resistivity 1 x 109 ± 10%
    Thickness (Metric) 2mm
  • Description & Specifications
Related Products
Product Content Feedback